siRNA Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003809.2 | NP_003800.1 | 7 | 1142 | ||
| NR_037146.1 | 8 | 1381 | ||||
| GenBank mRNA | AF030099.1 | - | 7 | 1063 | ||
| BC019047.2 | - | 8 | 1381 | 4766071 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFSF12| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003809.2 | NP_003800.1 | 7 | 1142 | ||
| NR_037146.1 | 8 | 1381 | ||||
| GenBank mRNA | AF030099.1 | - | 7 | 1063 | ||
| BC019047.2 | - | 8 | 1381 | 4766071 | ||