siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271757.1 | NP_001258686.1 | 3 | 433 | ||
| NM_001271758.1 | NP_001258687.1 | 3 | 376 | |||
| NM_009525.3 | NP_033551.2 | 3 | 630 | |||
| GenBank mRNA | AK013218.1 | - | 3 | 601 | ||
| AK049558.1 | - | 3 | 400 | |||
| BC010775.1 | - | 3 | 376 | 4211437 | ||
| BC051406.1 | - | 3 | 561 | 6438917 | ||
| M89799.1 | - | 2 | 245 | |||

Wnt5b
Genomic Map