siRNA Details
| Gene Symbol: Xpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009531.2 | NP_033557.2 | 2 | 352 | ||
| GenBank mRNA | AB071144.1 | - | 2 | 289 | ||
| AK004713.1 | - | 2 | 292 | |||
| AK028595.1 | - | 2 | 290 | |||
| AK166981.1 | - | 2 | 287 | |||
| AK170975.1 | - | 2 | 284 | |||
| AK191690.1 | - | 1 | 172 | |||
| BC052725.1 | - | 2 | 314 | 6812198 | ||
| BY238377.1 | - | 2 | 353 | |||
| U27398.1 | - | 2 | 260 | |||
| U40005.1 | - | 1 | 91 | |||

Xpc
Genomic Map