siRNA Details
| Gene Symbol: FOXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004474.3 | NP_004465.3 | 1 | 3693 | ||
| GenBank mRNA | AF042832.1 | - | 1 | 3707 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXD2| Gene Symbol: FOXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004474.3 | NP_004465.3 | 1 | 3693 | ||
| GenBank mRNA | AF042832.1 | - | 1 | 3707 | ||