siRNA Details
| Gene Symbol: Tuba3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009449.3 | NP_033475.1 | 3 | 468 | ||
| GenBank mRNA | BC050769.1 | - | 3 | 392 | 6771187 | |
| CK137160.1 | - | 3 | 468 | |||
| M13443.1 | - | 3 | 383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tuba3b| Gene Symbol: Tuba3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009449.3 | NP_033475.1 | 3 | 468 | ||
| GenBank mRNA | BC050769.1 | - | 3 | 392 | 6771187 | |
| CK137160.1 | - | 3 | 468 | |||
| M13443.1 | - | 3 | 383 | |||