siRNA Details
| Gene Symbol: Ubr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009461.2 | NP_033487.2 | 2 | 424 | ||
| GenBank mRNA | AF061555.1 | - | 2 | 425 | ||
| AK089616.1 | - | 2 | 324 | |||
| AK132284.1 | - | 2 | 338 | |||
| AK158467.1 | - | 402 | ||||
| AK162013.1 | - | 2 | 325 | |||
| AK166885.1 | - | 2 | 322 | |||
| BC090969.1 | - | 2 | 312 | |||
| BC100461.1 | - | 2 | 331 | |||

Ubr1
Genomic Map