siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009516.3 | NP_033542.2 | 4 | 1252 | ||
| GenBank mRNA | AK011212.1 | - | 4 | 1028 | ||
| AK039572.1 | - | 4 | 568 | |||
| BC006852.1 | - | 4 | 1271 | 3156953 | ||
| D30743.1 | - | 4 | 1253 | |||

Wee1
Genomic Map