siRNA Details
| Gene Symbol: Tspyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009433.3 | NP_033459.1 | 1 | 939 | ||
| GenBank mRNA | AF042180.1 | - | 1 | 903 | ||
| AK053812.1 | - | 1 | 890 | |||
| AK159432.1 | - | 1 | 920 | |||
| AK166438.1 | - | 1 | 920 | |||
| AK166805.1 | - | 1 | 868 | |||
| AK166893.1 | - | 1 | 868 | |||
| BC011213.1 | - | 1 | 886 | 4196841 | ||

Tspyl1
Genomic Map