siRNA Details
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009441.2 | NP_033467.2 | 33 | 3731 | ||
GenBank mRNA | AB008516.1 | - | 32 | 3608 | ||
AK148494.1 | - | 6 | 755 | |||
AK161168.1 | - | 1 | 2644 | |||
AK164492.1 | - | 1 | 253 | |||
BC145489.1 | - | 28 | 3251 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
