siRNA Details
| Gene Symbol: Ube2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169576.1 | NP_001163047.1 | 5 | 1235 | ||
| NM_001169577.1 | NP_001163048.1 | 5 | 811 | |||
| NM_009459.3 | NP_033485.1 | 7 | 1328 | |||
| GenBank mRNA | BC008517.1 | - | 7 | 1134 | 3600233 | |
| U19854.1 | - | 7 | 1057 | |||

Ube2h
Genomic Map