siRNA Details
| Gene Symbol: Ubr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009461.2 | NP_033487.2 | 27 | 3046 | ||
| GenBank mRNA | AF061555.1 | - | 27 | 3047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr1| Gene Symbol: Ubr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009461.2 | NP_033487.2 | 27 | 3046 | ||
| GenBank mRNA | AF061555.1 | - | 27 | 3047 | ||