siRNA Details
| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293561.1 | NP_001280490.1 | 17 | 5497 | ||
| NM_009472.4 | NP_033498.1 | 16 | 5440 | |||
| GenBank mRNA | AK141130.1 | - | 1 | 1839 | ||
| U72634.1 | - | 16 | 5441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc5c| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293561.1 | NP_001280490.1 | 17 | 5497 | ||
| NM_009472.4 | NP_033498.1 | 16 | 5440 | |||
| GenBank mRNA | AK141130.1 | - | 1 | 1839 | ||
| U72634.1 | - | 16 | 5441 | |||