siRNA Details
| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009474.5 | NP_033500.1 | 8 | 1043 | ||
| GenBank mRNA | AK050047.1 | - | 7 | 833 | ||
| AK050425.1 | - | 8 | 1044 | |||
| BC019771.1 | - | 8 | 1025 | 5136328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uox| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009474.5 | NP_033500.1 | 8 | 1043 | ||
| GenBank mRNA | AK050047.1 | - | 7 | 833 | ||
| AK050425.1 | - | 8 | 1044 | |||
| BC019771.1 | - | 8 | 1025 | 5136328 | ||