siRNA Details
| Gene Symbol: Upk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009476.2 | NP_033502.2 | 3, 4 | 403 | ||
| GenBank mRNA | BC104260.1 | - | 3, 4 | 403 | 40039813 | |
| BC104261.1 | - | 3, 4 | 403 | 40039814 | ||
| DQ358978.1 | - | 3, 4 | 342 | |||
| U08030.1 | - | 3, 4 | 401 | |||

Upk2
Genomic Map