siRNA Details
| Gene Symbol: Uty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009484.2 | NP_033510.2 | 15 | 1799 | ||
| GenBank mRNA | AF057367.1 | - | 15 | 1779 | ||
| AK142344.1 | - | 18 | 1937 | |||
| AK142512.1 | - | 15 | 1715 | |||
| AK155922.1 | - | 15 | 1544 | |||
| AK162410.1 | - | 13 | 1372 | |||
| AK164380.1 | - | 1 | 1736 | |||
| AK164917.1 | - | 2 | 244 | |||
| AK220463.1 | - | 16 | 1756 | |||
| BC053061.1 | - | 15 | 1782 | |||

Uty
Genomic Map