siRNA Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 6 | 2497 | ||
| GenBank mRNA | AF012923.1 | - | 7 | 2386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmat3| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 6 | 2497 | ||
| GenBank mRNA | AF012923.1 | - | 7 | 2386 | ||