siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271757.1 | NP_001258686.1 | 5 | 1625 | ||
| NM_001271758.1 | NP_001258687.1 | 5 | 1568 | |||
| NM_009525.3 | NP_033551.2 | 5 | 1822 | |||
| GenBank mRNA | AK013218.1 | - | 5 | 1793 | ||
| AK049558.1 | - | 5 | 1593 | |||
| BC010775.1 | - | 5 | 1568 | 4211437 | ||
| BC051406.1 | - | 5 | 1753 | 6438917 | ||
| M89799.1 | - | 4 | 1433 | |||

Wnt5b
Genomic Map