siRNA Details
| Gene Symbol: Wnt7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009527.3 | NP_033553.2 | 4 | 2121 | ||
| GenBank mRNA | AK004683.1 | - | 4 | 2122 | ||
| BC049093.1 | - | 4 | 2115 | 6415801 | ||
| BC057586.1 | - | 4 | 2115 | 6415801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt7a| Gene Symbol: Wnt7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009527.3 | NP_033553.2 | 4 | 2121 | ||
| GenBank mRNA | AK004683.1 | - | 4 | 2122 | ||
| BC049093.1 | - | 4 | 2115 | 6415801 | ||
| BC057586.1 | - | 4 | 2115 | 6415801 | ||