siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 9 | 2130 | ||
| GenBank mRNA | AF026032.1 | - | 10 | 2097 | ||
| AK032955.1 | - | 5 | 1542 | |||
| AK080333.1 | - | 1 | 392 | |||
| X99643.1 | - | 1 | 905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atrx| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 9 | 2130 | ||
| GenBank mRNA | AF026032.1 | - | 10 | 2097 | ||
| AK032955.1 | - | 5 | 1542 | |||
| AK080333.1 | - | 1 | 392 | |||
| X99643.1 | - | 1 | 905 | |||