siRNA Details
| Gene Symbol: Zfp62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024846.1 | NP_001020017.1 | 5 | 386 | ||
| NM_009562.2 | NP_033588.2 | 4 | 286 | |||
| GenBank mRNA | AK028351.1 | - | 5 | 389 | ||
| AK048619.1 | - | 4 | 288 | |||
| AK088060.1 | - | 4 | 464 | |||
| AK154767.1 | - | 4 | 277 | |||
| AK157730.1 | - | 4 | 370 | |||
| AK165654.1 | - | 4 | 268 | |||
| AK167091.1 | - | 4 | 301 | |||
| BC022935.1 | - | 4 | 404 | 3497847 | ||
| BC065692.1 | - | 2 | 490 | 6826605 | ||

Zfp62
Genomic Map