siRNA Details
| Gene Symbol: Zfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009571.2 | NP_033597.2 | 6 | 1196 | ||
| GenBank mRNA | AK030048.1 | - | 6 | 1067 | ||
| AK133222.1 | - | 6 | 1197 | |||
| M24401.1 | - | 6 | 1157 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfy2| Gene Symbol: Zfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009571.2 | NP_033597.2 | 6 | 1196 | ||
| GenBank mRNA | AK030048.1 | - | 6 | 1067 | ||
| AK133222.1 | - | 6 | 1197 | |||
| M24401.1 | - | 6 | 1157 | |||