siRNA Details
| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009632.2 | NP_033762.1 | 2 | 163 | ||
| GenBank mRNA | AF072521.1 | - | 2 | 138 | ||
| AJ007780.1 | - | 2 | 176 | |||
| AK076193.1 | - | 2 | 190 | |||
| AK141876.1 | - | 2 | 185 | |||
| BC052000.1 | - | 2 | 163 | |||
| BC062150.1 | - | 2 | 163 | 6333875 | ||

Parp2
Genomic Map