siRNA Details
| Gene Symbol: Yy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009537.3 | NP_033563.2 | 4 | 1033 | ||
| GenBank mRNA | AK076149.1 | - | 5 | 646 | ||
| AK166542.1 | - | 5 | 371 | |||
| BC055899.1 | - | 4 | 1011 | 3156776 | ||
| M73963.1 | - | 4 | 1033 | |||
| M74590.1 | - | 4 | 973 | |||

Yy1
Genomic Map