siRNA Details
| Gene Symbol: GPR160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014373.2 | NP_055188.1 | 4 | 1093 | ||
| XM_005247346.3 | XP_005247403.1 | 5 | 1703 | |||
| XM_005247347.3 | XP_005247404.1 | 3 | 739 | |||
| XM_006713582.3 | XP_006713645.1 | 4 | 1055 | |||
| XM_011512678.2 | XP_011510980.1 | 5 | 1385 | |||
| XM_017006161.1 | XP_016861650.1 | 5 | 1196 | |||
| XM_017006162.1 | XP_016861651.1 | 5 | 1200 | |||
| XM_017006163.1 | XP_016861652.1 | 4 | 1717 | |||
| GenBank mRNA | AJ249248.1 | - | 3 | 824 | ||
| BC000181.2 | - | 4 | 1028 | 3048193 | ||

GPR160
Genomic Map