siRNA Details
| Gene Symbol: Zfp101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009542.2 | NP_033568.2 | 4 | 542 | ||
| GenBank mRNA | AK076355.1 | - | 4 | 546 | ||
| AK076461.1 | - | 1 | 332 | |||
| AK078458.1 | - | 4 | 530 | |||
| AK169234.1 | - | 4 | 543 | |||
| U07861.1 | - | 4 | 502 | |||

Zfp101
Genomic Map