siRNA Details
| Gene Symbol: Zfp60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009560.2 | NP_033586.2 | 5 | 555 | ||
| NM_029531.2 | NP_083807.2 | 5 | 361 | |||
| GenBank mRNA | AA718695.1 | - | 5 | 361 | ||
| AK028717.1 | - | 5 | 455 | |||
| AK149305.1 | - | 5 | 382 | |||
| BC031754.1 | - | 5 | 360 | 3992720 | ||
| CB526626.1 | - | 5 | 498 | |||

Zfp60
Genomic Map