siRNA Details
| Gene Symbol: Zfp93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009567.4 | NP_033593.1 | 4 | 774 | ||
| GenBank mRNA | AK028905.1 | - | 4 | 612 | ||
| AK040325.1 | - | 4 | 705 | |||
| AK132229.1 | - | 4 | 775 | |||
| AK146095.1 | - | 4 | 765 | |||
| BC003776.1 | - | 497 | 3496816 | |||
| BC061487.1 | - | 3 | 555 | 5349153 | ||
| U46186.1 | - | 4 | 766 | |||

Zfp93
Genomic Map