siRNA Details
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009569.3 | NP_033595.1 | 3 | 617 | ||
GenBank mRNA | AF006492.1 | - | 3 | 621 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009569.3 | NP_033595.1 | 3 | 617 | ||
GenBank mRNA | AF006492.1 | - | 3 | 621 |