siRNA Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009593.2 | NP_033723.1 | 2 | 369 | ||
| GenBank mRNA | AF323659.1 | - | 2 | 365 | ||
| AK037433.1 | - | 2 | 289 | |||
| BC119471.2 | - | 2 | 317 | 40048166 | ||
| U34920.1 | - | 2 | 327 | |||
| Z48745.1 | - | 2 | 356 | |||

Abcg1
Genomic Map