siRNA Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF053943.1 | - | 2 | 493 | ||
| AK158276.1 | - | 2 | 517 | |||
| AK159330.1 | - | 2 | 497 | |||
| AK159342.1 | - | 2 | 516 | |||
| AK159377.1 | - | 2 | 487 | |||
| AK159409.1 | - | 2 | 517 | |||
| AK159774.1 | - | 2 | 488 | |||
| AK159957.1 | - | 2 | 488 | |||
| AK189675.1 | - | 2 | 60 | |||
| AK200329.1 | - | 2 | 60 | |||
| BC082577.1 | - | 2 | 505 | 30648530 | ||

Aebp1
Genomic Map