siRNA Details
| Gene Symbol: BCAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008405.2 | NP_001008405.1 | 3 | 234 | ||
| NM_018844.3 | NP_061332.2 | 4 | 559 | |||
| NR_027830.1 | 3 | 453 | ||||
| XM_006716051.2 | XP_006716114.1 | 4 | 555 | |||
| XM_011516406.2 | XP_011514708.1 | 4 | 318 | |||
| XM_011516407.1 | XP_011514709.1 | 4 | 555 | |||
| XM_011516408.2 | XP_011514710.1 | 4 | 555 | |||
| XM_011516409.2 | XP_011514711.1 | 4 | 555 | |||
| XM_017012436.1 | XP_016867925.1 | 4 | 555 | |||
| XM_017012437.1 | XP_016867926.1 | 4 | 555 | |||
| XM_017012438.1 | XP_016867927.1 | 4 | 555 | |||
| GenBank mRNA | AF126020.1 | - | 4 | 266 | ||
| AK289401.1 | - | 4 | 316 | |||
| AK294824.1 | - | 3 | 184 | |||
| BC008478.1 | - | 4 | 271 | 4294273 | ||
| BG776708.1 | - | 4 | 560 | |||
| BM023571.1 | - | 1 | 29 | |||
| BT006981.1 | - | 3 | 220 | |||
| BX647653.1 | - | 4 | 270 | |||
| CK820004.1 | - | 2 | 91 | |||

BCAP29
Genomic Map