siRNA Details
| Gene Symbol: Zfp93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009567.4 | NP_033593.1 | 4 | 1828 | ||
| GenBank mRNA | AK028905.1 | - | 4 | 1666 | ||
| AK040325.1 | - | 4 | 1759 | |||
| AK132229.1 | - | 4 | 1829 | |||
| AK146095.1 | - | 4 | 1819 | |||
| AK165630.1 | - | 4 | 1783 | |||
| BC003776.1 | - | 1551 | 3496816 | |||
| BC061487.1 | - | 3 | 1609 | 5349153 | ||
| U46186.1 | - | 4 | 1820 | |||

Zfp93
Genomic Map