siRNA Details
| Gene Symbol: Zfp94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199321.1 | NP_001186250.1 | 6 | 1878 | ||
| NM_009568.3 | NP_033594.1 | 5 | 1839 | |||
| GenBank mRNA | AK138205.1 | - | 6 | 1819 | ||
| AK164334.1 | - | 7 | 1327 | |||
| BC005437.1 | - | 5 | 1839 | 3591835 | ||
| BC095945.1 | - | 4 | 1659 | 4936327 | ||
| U46187.1 | - | 5 | 1782 | |||

Zfp94
Genomic Map