siRNA Details
| Gene Symbol: Zik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009577.3 | NP_033603.2 | 4 | 2069 | ||
| GenBank mRNA | AK146692.1 | - | 4 | 2063 | ||
| AK161888.1 | - | 1 | 481 | |||
| BC050942.1 | - | 4 | 2027 | 6821378 | ||
| BC067196.1 | - | 4 | 2027 | 6821378 | ||

Zik1
Genomic Map