siRNA Details
| Gene Symbol: Zp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009580.2 | NP_033606.2 | 12 | 1892 | ||
| GenBank mRNA | BC100347.1 | - | 12 | 1910 | ||
| BC125613.1 | - | 12 | 1892 | 40130128 | ||
| BC125615.1 | - | 12 | 1892 | 40130130 | ||
| U20448.1 | - | 12 | 1923 | |||

Zp1
Genomic Map