siRNA Details
| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009671.5 | NP_033801.4 | 2 | 160 | ||
| GenBank mRNA | AB011370.2 | - | 2 | 158 | ||
| AB098329.1 | - | 2 | 158 | |||
| AK134008.1 | - | 2 | 157 | |||
| BC139231.1 | - | 2 | 171 | 8862253 | ||
| BC145583.1 | - | 2 | 171 | 9054149 | ||
| BQ550282.1 | - | 2 | 165 | |||

Ankfy1
Genomic Map