siRNA Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110300.1 | NP_001103770.1 | 2 | 160 | ||
| NM_009678.2 | NP_033808.2 | 2 | 160 | |||
| GenBank mRNA | AF067146.1 | - | 2 | 134 | ||
| AK010478.1 | - | 2 | 139 | |||
| AK082674.1 | - | 144 | ||||
| AK160161.1 | - | 2 | 127 | |||
| BC003704.1 | - | 2 | 99 | 3488231 | ||
| CJ078558.1 | - | 2 | 161 | |||

Ap1m2
Genomic Map