siRNA Details
| Gene Symbol: Bcl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009745.2 | NP_033875.2 | 2 | 274 | ||
| GenBank mRNA | AJ011145.1 | - | 2 | 154 | ||
| AK131643.1 | - | 2 | 170 | |||
| AK155076.1 | - | 2 | 156 | |||
| AK160439.1 | - | 2 | 257 | |||
| AK186334.1 | - | 2 | 103 | |||
| BC011471.1 | - | 2 | 251 | 3660754 | ||
| BY087167.1 | - | 2 | 274 | |||

Bcl7b
Genomic Map