siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 4 | 383 | ||
| GenBank mRNA | AB221611.1 | - | 3 | 177 | ||
| AK086173.1 | - | 4 | 385 | |||
| AK144702.1 | - | 4 | 302 | |||
| BB662609.1 | - | 4 | 385 | |||
| BC068303.1 | - | 4 | 219 | 30431022 | ||
| U31625.1 | - | 4 | 240 | |||
| U32446.1 | - | 4 | 293 | |||
| U35641.1 | - | 4 | 276 | |||
| U36475.1 | - | 3 | 203 | |||
| U68174.1 | - | 4 | 287 | |||

Brca1
Genomic Map