siRNA Details
| Gene Symbol: Ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF084575.1 | - | 2, 3 | 238 | ||
| AK002565.1 | - | 2, 3 | 264 | |||
| AK137489.1 | - | 2, 3 | 259 | |||
| AK152100.1 | - | 2, 3 | 270 | |||
| AK194047.1 | - | 20 | ||||
| BC012656.1 | - | 2, 3 | 249 | 4187607 | ||
| BC108327.1 | - | 2, 3 | 257 | 30713247 | ||
| BC131985.1 | - | 2, 3 | 239 | 40130262 | ||
| BC145234.1 | - | 2, 3 | 239 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106933, XM_006254693, XM_006254694, XM_008772548, XM_006254695, and others.

Ap3s1
Genomic Map