siRNA Details
| Gene Symbol: Cd5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009690.2 | NP_033820.2 | 4 | 634 | ||
| GenBank mRNA | AF011428.1 | - | 4 | 540 | ||
| AF018268.1 | - | 4 | 573 | |||
| AF018269.1 | - | 4 | 559 | |||
| AK159132.1 | - | 4 | 583 | |||
| AK159181.1 | - | 4 | 637 | |||
| AK159823.1 | - | 4 | 602 | |||
| BC006799.1 | - | 4 | 575 | 3152586 | ||
| BC094459.1 | - | 4 | 587 | 4161650 | ||

Cd5l
Genomic Map