siRNA Details
| Gene Symbol: Bcl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009745.2 | NP_033875.2 | 3, 4 | 407 | ||
| GenBank mRNA | AJ011145.1 | - | 3, 4 | 287 | ||
| AK131643.1 | - | 3, 4 | 303 | |||
| AK155076.1 | - | 3, 4 | 289 | |||
| AK186334.1 | - | 3, 4 | 236 | |||
| BC011471.1 | - | 3, 4 | 384 | 3660754 | ||

Bcl7b
Genomic Map