siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 5 | 436 | ||
| GenBank mRNA | AB221611.1 | - | 4 | 230 | ||
| AK144702.1 | - | 5 | 355 | |||
| BC068303.1 | - | 5 | 272 | 30431022 | ||
| U31625.1 | - | 5 | 293 | |||
| U32446.1 | - | 5 | 346 | |||
| U35641.1 | - | 5 | 329 | |||
| U36475.1 | - | 4 | 256 | |||
| U68174.1 | - | 5 | 340 | |||

Brca1
Genomic Map