siRNA Details
| Gene Symbol: Ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF084575.1 | - | 2, 3 | 241 | ||
| AK002565.1 | - | 2, 3 | 267 | |||
| AK137489.1 | - | 2, 3 | 262 | |||
| AK152100.1 | - | 2, 3 | 273 | |||
| AK194047.1 | - | 23 | ||||
| BC012656.1 | - | 2, 3 | 252 | 4187607 | ||
| BC108327.1 | - | 2, 3 | 260 | 30713247 | ||
| BC131985.1 | - | 2, 3 | 242 | 40130262 | ||
| BC145234.1 | - | 2, 3 | 242 | |||

Ap3s1
Genomic Map