siRNA Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135254.1 | NP_001128726.1 | 4 | 1116 | ||
| NM_002584.2 | NP_002575.1 | 4 | 1116 | |||
| NM_013945.2 | NP_039236.1 | 4 | 1110 | |||
| GenBank mRNA | BC121165.2 | - | 4 | 592 | 40121582 | |
| BC121166.2 | - | 4 | 592 | 40121584 | ||
| DQ322591.1 | - | 4 | 601 | |||
| X96743.1 | - | 4 | 1117 | |||
| Z35141.1 | - | 4 | 620 | |||

PAX7
Genomic Map