siRNA Details
| Gene Symbol: Apoe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010261.1 | - | 4 | 998 | ||
| AK019319.1 | - | 1 | 733 | |||
| AK131624.1 | - | 4 | 998 | |||
| AK134921.1 | - | 4 | 998 | |||
| AK149568.1 | - | 4 | 980 | |||
| AK150834.1 | - | 4 | 1000 | |||
| AK159105.1 | - | 4 | 998 | |||
| AK159424.1 | - | 4 | 1000 | |||
| AK159517.1 | - | 4 | 998 | |||
| AK197774.1 | - | 1 | 125 | |||
| BC028816.1 | - | 4 | 977 | 5136415 | ||
| BC044785.1 | - | 3 | 969 | |||
| BC083351.1 | - | 3 | 967 | 6818357 | ||
| M12414.1 | - | 3 | 932 | |||
| M73490.1 | - | 2 | 859 | |||

Apoe
Genomic Map