siRNA Details
| Gene Symbol: RING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002931.3 | NP_002922.2 | 5, 6 | 1047 | ||
| GenBank mRNA | AK289355.1 | - | 5, 6 | 1056 | ||
| AK301378.1 | - | 5, 6 | 990 | |||
| BC002922.2 | - | 5, 6 | 1042 | 3954313 | ||
| BC051866.2 | - | 5, 6 | 1040 | 6142438 | ||
| BT007352.1 | - | 3, 4 | 766 | |||
| Z14000.1 | - | 3, 4 | 841 | |||

RING1
Genomic Map