siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 10 | 4036 | ||
| GenBank mRNA | AB221610.1 | - | 1 | 1265 | ||
| BC068303.1 | - | 10 | 3872 | 30431022 | ||
| U31625.1 | - | 10 | 3893 | |||
| U32446.1 | - | 10 | 3946 | |||
| U35641.1 | - | 10 | 3929 | |||
| U36475.1 | - | 9 | 3856 | |||
| U68174.1 | - | 10 | 3940 | |||

Brca1
Genomic Map