siRNA Details
| Gene Symbol: C3ar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009779.2 | NP_033909.1 | 2 | 3056 | ||
| GenBank mRNA | AK080984.1 | - | 1 | 416 | ||
| AK081010.1 | - | 1383 | ||||
| U77460.1 | - | 2 | 2904 | |||
More Information
Set Membership: |
|

C3ar1
Genomic Map