siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 3, 4 | 320 | ||
| AK029512.1 | - | 3, 4 | 366 | |||
| AK030601.1 | - | 3, 4 | 346 | |||
| BC011278.1 | - | 3, 4 | 331 | 3674983 | ||
| CN696192.1 | - | 3, 4 | 369 | |||

Entpd1
Genomic Map